Ion chromatography standard, lithium, VHG
Lieferant: LGC Standards PROMOCHEM
VHG-ILI100-500
VHG-ILIW1K-100
PROOVHG-ILI100-500EA
0
EUR
PROOVHG-ILI100-500
PROOVHG-ILIW1K-100
Ion chromatography standard, lithium, VHG
Standards
Kationischer Standards
Lithium-Standards
This solution is intended for use as a certified reference material and calibration standard, for ion chromatography (IC) and techniques using other modes of aqueous ion detection.
Standards: Traceable to NIST and SRM.