Nitrate(N), ion chromatography standard, VHG
Lieferant: LGC Standards PROMOCHEM
VHG-INO3N-500
VHG-INO3N-100
PROOVHG-INO3N-500EA
110
EUR
PROOVHG-INO3N-500
PROOVHG-INO3N-100
Nitrate(N), ion chromatography standard, VHG
Standards
Anionische Standards
Nitrat (N)-Standards
This solution is intended for use as a certified reference material (CRM) and calibration standard, for ion chromatography (IC) and techniques using other modes of aqueous ion detection.
Standards: Traceable to NIST and SRM.