Phosphate ion chromatography standard (PO₄⁻³ as P), VHG
Lieferant: LGC Standards PROMOCHEM
VHG-IPO4P-100 VHG-IPO4P-500
PROOVHG-IPO4P-100EA 58 EUR
PROOVHG-IPO4P-100 PROOVHG-IPO4P-500
Phosphate ion chromatography standard (PO₄⁻³ as P), VHG
Standards Anionische Standards Phosphat (P)-Standards
This solution is intended for use as a certified reference material (CRM) and calibration standard, for ion chromatography (IC) and techniques using other modes of aqueous ion detection.
Standards: Traceable to NIST and SRM.
Bestellen
TECHNISCHE DATEN
Environmentally Preferable