Lithium isotopic standard solution, for ICP, CRM, VHG
Lieferant: LGC Standards PROMOCHEM
VHG-LIS6LIZ-10
PROOVHG-LIS6LIZ-10EA
116
EUR
PROOVHG-LIS6LIZ-10
Lithium isotopic standard solution, for ICP, CRM, VHG
Standards
Kationischer Standards
Lithium-Standards
This solution is intended for use as a calibration or reference standard for inductively coupled plasma mass spectrometry (ICP-MS) where known values for isotopic abundance are required.
Standards: Traceable to NIST, SRM.