Selenium standard solution, for ICP, in HNO₃, CRM, VHG
Lieferant: LGC Standards PROMOCHEM
VHG-PSEN-100 VHG-LSEN-100 VHG-PSEN-500 VHG-TSEN-100 VHG-TSEN-500
PROOVHG-PSEN-100EA 50 EUR
PROOVHG-PSEN-100 PROOVHG-LSEN-100 PROOVHG-PSEN-500 PROOVHG-TSEN-100 PROOVHG-TSEN-500
Selenium standard solution, for ICP, in HNO₃, CRM, VHG
Standards Kationischer Standards Selen-Standards
These solutions are intended for use as a certified reference material or calibration standard for inductively coupled plasma optical emission spectroscopy (ICP-OES), inductively coupled plasma mass spectrometry (ICP-MS), flame or furnace atomic absorption spectroscopy (AA or GFAA), x-ray fluorescence spectroscopy (XRF), and other techniques for elemental analysis.
Standards: Traceable to NIST, SRM.