Single element petroleum standard, sulphur free lead in hydrocarbon oil, CRM, VHG
Lieferant: LGC Standards PROMOCHEM
VHG-OSF-PB1000
PROOVHG-OSF-PB1000EA 0 EUR
PROOVHG-OSF-PB1000
Single element petroleum standard, sulphur free lead in hydrocarbon oil, CRM, VHG
Standards Kationischer Standards Blei-Standards
This solution is intended for use as a certified reference material or calibration standard for the analysis of this element in petroleum products or other organic matrices using x-ray fluorescence (XRF) spectrometry, inductively coupled plasma optical emission spectroscopy (ICPOES), inductively coupled plasma mass spectrometry (ICP-MS), rotating disc electrode atomic emission spectroscopy (RDE-AES), flame or furnace atomic absorption spectroscopy (AA or GFAA), and other techniques for elemental analysis.
Standards: Traceable to NIST, SRM.