Arsenic standard solution, for ICP, in 5% HNO₃, VHG
Lieferant: LGC Standards PROMOCHEM
VHG-PASN-100 VHG-PASN-500 VHG-TASN-100 VHG-TASN-500
PROOVHG-PASN-100EA 57 EUR
PROOVHG-PASN-100 PROOVHG-PASN-500 PROOVHG-TASN-100 PROOVHG-TASN-500
Arsenic standard solution, for ICP, in 5% HNO₃, VHG
Standards Kationischer Standards Arsen-Standards
The solutions are intended for use as a certified reference material or calibration standard for inductively coupled plasma optical emission spectroscopy (ICP-OES), inductively coupled plasma mass spectrometry (ICP-MS), flame or furnace atomic absorption spectroscopy (AA or GFAA), x-ray fluorescence spectroscopy (XRF), and other techniques for elemental analysis.
Standards: Traceable to NIST, SRM.