Bismuth standard solution, for ICP, in 5% HNO₃, VHG
Lieferant: LGC Standards PROMOCHEM
VHG-PBIN-100 VHG-PBIN-500 VHG-TBIN-100 VHG-TBIN-500
PROOVHG-PBIN-100EA 50 EUR
PROOVHG-PBIN-100 PROOVHG-PBIN-500 PROOVHG-TBIN-100 PROOVHG-TBIN-500
Bismuth standard solution, for ICP, in 5% HNO₃, VHG
Standards Kationischer Standards Wismut-Standards
The solutions are intended for use as a certified reference material or calibration standard for inductively coupled plasma optical emission spectroscopy (ICP-OES), inductively coupled plasma mass spectrometry (ICP-MS), flame or furnace atomic absorption spectroscopy (AA or GFAA), x-ray fluorescence spectroscopy (XRF), and other techniques for elemental analysis.
Standards: Traceable to NIST, SRM.