Calcium standard solution, for ICP, in 5% HNO₃, VHG
Lieferant: LGC Standards PROMOCHEM
VHG-PCAN-100 VHG-TCAN-100 VHG-TCAN-500 VHG-PCAN-50 VHG-PCAN-500
PROOVHG-PCAN-100EA 50 EUR
PROOVHG-PCAN-100 PROOVHG-TCAN-100 PROOVHG-TCAN-500 PROOVHG-PCAN-50 PROOVHG-PCAN-500
Calcium standard solution, for ICP, in 5% HNO₃, VHG
Standards Kationischer Standards Calcium-Standards
The solutions are intended for use as a certified reference material or calibration standard for inductively coupled plasma optical emission spectroscopy (ICP-OES), inductively coupled plasma mass spectrometry (ICP-MS), flame or furnace atomic absorption spectroscopy (AA or GFAA), x-ray fluorescence spectroscopy (XRF), and other techniques for elemental analysis.
Standards: Traceable to NIST, SRM.