Hexavalent chromium standard solution, in H?O, VHG
Lieferant: LGC Standards PROMOCHEM
VHG-PCR6W-100 VHG-PCR6W-50 VHG-PCR6W-500
PROOVHG-PCR6W-100EA 51 EUR
PROOVHG-PCR6W-100 PROOVHG-PCR6W-50 PROOVHG-PCR6W-500
Hexavalent chromium standard solution, in H?O, VHG
Standards Kationischer Standards Chrom-Standards
The solutions are intended for use as a certified reference material or calibration standard for inductively coupled plasma optical emission spectroscopy (ICP-OES), inductively coupled plasma mass spectrometry (ICP-MS), flame or furnace atomic absorption spectroscopy (AA or GFAA), x-ray fluorescence spectroscopy (XRF), and other techniques for elemental analysis.
Standards: Traceable to NIST, SRM.