Copper standard solution, for ICP, in 5% HNO₃, VHG
Lieferant: LGC Standards PROMOCHEM
VHG-PCUN-100
VHG-PCUN-500
VHG-PCUN-250
VHG-PCUN-50
VHG-TCUN-100
VHG-TCUN-500
PROOVHG-PCUN-100EA
50
EUR
PROOVHG-PCUN-100
PROOVHG-PCUN-500
PROOVHG-PCUN-250
PROOVHG-PCUN-50
PROOVHG-TCUN-100
PROOVHG-TCUN-500
Copper standard solution, for ICP, in 5% HNO₃, VHG
Standards
Kationischer Standards
Kupfer-Standards
The solutions are intended for use as a certified reference material or calibration standard for inductively coupled plasma optical emission spectroscopy (ICP-OES), inductively coupled plasma mass spectrometry (ICP-MS), flame or furnace atomic absorption spectroscopy (AA or GFAA), x-ray fluorescence spectroscopy (XRF), and other techniques for elemental analysis.
Standards: Traceable to NIST, SRM.