Iron standard solution, for ICP, in 5% HNO₃, VHG
Lieferant: LGC Standards PROMOCHEM
VHG-PFEN-100
VHG-PFEN-500
VHG-TFEN-100
VHG-PFEN-50
VHG-TFEN-500
PROOVHG-PFEN-100EA
50
EUR
PROOVHG-PFEN-100
PROOVHG-PFEN-500
PROOVHG-TFEN-100
PROOVHG-PFEN-50
PROOVHG-TFEN-500
Iron standard solution, for ICP, in 5% HNO₃, VHG
Standards
Kationischer Standards
Eisen-Standards
The solutions are intended for use as a certified reference material or calibration standard for inductively coupled plasma optical emission spectroscopy (ICP-OES), inductively coupled plasma mass spectrometry (ICP-MS), flame or furnace atomic absorption spectroscopy (AA or GFAA), x-ray fluorescence spectroscopy (XRF), and other techniques for elemental analysis.
Standards: Traceable to NIST, SRM.