Rhenium standard solution, for ICP, in 5% HNO₃, VHG
Lieferant: LGC Standards PROMOCHEM
VHG-PREN-100 VHG-PREN-50 VHG-PREN-500 VHG-TREN-100 VHG-TREN-500
PROOVHG-PREN-100EA 102 EUR
PROOVHG-PREN-100 PROOVHG-PREN-50 PROOVHG-PREN-500 PROOVHG-TREN-100 PROOVHG-TREN-500
Rhenium standard solution, for ICP, in 5% HNO₃, VHG
Standards Kationischer Standards Rhenium-Standards
The solutions are intended for use as a certified reference material or calibration standard for inductively coupled plasma optical emission spectroscopy (ICP-OES), inductively coupled plasma mass spectrometry (ICP-MS), flame or furnace atomic absorption spectroscopy (AA or GFAA), x-ray fluorescence spectroscopy (XRF), and other techniques for elemental analysis.
Standards: Traceable to NIST, SRM.