Rhodium standard solution, for ICP, in 20% HCl, VHG
Lieferant: LGC Standards PROMOCHEM
VHG-PRHH-100
VHG-PRHH-50
VHG-PRHH-500
VHG-TRHH-100
VHG-TRHH-500
PROOVHG-PRHH-100EA
581
EUR
PROOVHG-PRHH-100
PROOVHG-PRHH-50
PROOVHG-PRHH-500
PROOVHG-TRHH-100
PROOVHG-TRHH-500
Rhodium standard solution, for ICP, in 20% HCl, VHG
Standards
Kationischer Standards
Rhodium-Standards
The solutions are intended for use as a certified reference material or calibration standard for inductively coupled plasma optical emission spectroscopy (ICP-OES), inductively coupled plasma mass spectrometry (ICP-MS), flame or furnace atomic absorption spectroscopy (AA or GFAA), x-ray fluorescence spectroscopy (XRF), and other techniques for elemental analysis.
Standards: Traceable to NIST, SRM.