Strontium standard solution, for ICP, in HNO₃, CRM, VHG
Lieferant: LGC Standards PROMOCHEM
VHG-PSRN-100
VHG-PSRN-500
VHG-LSRN-100
VHG-PSRN-50
VHG-TSRN-100
VHG-TSRN-500
PROOVHG-PSRN-100EA
50
EUR
PROOVHG-PSRN-100
PROOVHG-PSRN-500
PROOVHG-LSRN-100
PROOVHG-PSRN-50
PROOVHG-TSRN-100
PROOVHG-TSRN-500
Strontium standard solution, for ICP, in HNO₃, CRM, VHG
Standards
Kationischer Standards
Strontium-Standards
These solutions are intended for use as a certified reference material or calibration standard for inductively coupled plasma optical emission spectroscopy (ICP-OES), inductively coupled plasma mass spectrometry (ICP-MS), flame or furnace atomic absorption spectroscopy (AA or GFAA), x-ray fluorescence spectroscopy (XRF), and other techniques for elemental analysis.
Standards: Traceable to NIST, SRM.