Arsenic standard solution, for ICP, in 5% HNO₃, VHG
Lieferant: LGC Standards PROMOCHEM
VHG-PASN-100
VHG-PASN-500
VHG-TASN-100
VHG-TASN-500
PROOVHG-PASN-100EA
57
EUR
PROOVHG-PASN-100
PROOVHG-PASN-500
PROOVHG-TASN-100
PROOVHG-TASN-500
Arsenic standard solution, for ICP, in 5% HNO₃, VHG
Standards
Kationischer Standards
Arsen-Standards
The solutions are intended for use as a certified reference material or calibration standard for inductively coupled plasma optical emission spectroscopy (ICP-OES), inductively coupled plasma mass spectrometry (ICP-MS), flame or furnace atomic absorption spectroscopy (AA or GFAA), x-ray fluorescence spectroscopy (XRF), and other techniques for elemental analysis.
Standards: Traceable to NIST, SRM.