Dysprosium standard solution, for ICP, in 5% HNO₃, CRM, VHG
Lieferant: LGC Standards PROMOCHEM
VHG-PDYN-100
VHG-PDYN-500
VHG-TDYN-100
VHG-TDYN-500
PROOVHG-PDYN-100EA
59
EUR
PROOVHG-PDYN-100
PROOVHG-PDYN-500
PROOVHG-TDYN-100
PROOVHG-TDYN-500
Dysprosium standard solution, for ICP, in 5% HNO₃, CRM, VHG
Standards
Kationischer Standards
Dysprosium-Standards
These solutions are intended for use as a certified reference material (CRM) or calibration standard for inductively coupled plasma optical emission spectroscopy (ICP-OES), inductively coupled plasma mass spectrometry (ICP-MS), flame or furnace atomic absorption spectroscopy (AA or GFAA), and other techniques for elemental analysis.
Standards: Traceable to NIST, SRM.