Europium standard solution, for ICP, in 5% HNO₃, CRM, VHG
Lieferant: LGC Standards PROMOCHEM
VHG-PEUN-100
VHG-PEUN-500
VHG-TEUN-100
VHG-TEUN-500
PROOVHG-PEUN-100EA
64
EUR
PROOVHG-PEUN-100
PROOVHG-PEUN-500
PROOVHG-TEUN-100
PROOVHG-TEUN-500
Europium standard solution, for ICP, in 5% HNO₃, CRM, VHG
Standards
Kationischer Standards
Europium-Standards
These solutions are intended for use as a certified reference material or calibration standard for inductively coupled plasma optical emission spectroscopy (ICP-OES), inductively coupled plasma mass spectrometry (ICP-MS), flame or furnace atomic absorption spectroscopy (AA or GFAA), x-ray fluorescence spectroscopy (XRF), and other techniques for elemental analysis.
Standards: Traceable to NIST, SRM.