Gallium standard solution, for ICP, in 5% HNO₃, 0,5% HCl, VHG
Lieferant: LGC Standards PROMOCHEM
VHG-PGANH-100 VHG-TGANH-100 VHG-PGANH-50 VHG-PGANH-500 VHG-TGANH-500
PROOVHG-PGANH-100EA 59 EUR
PROOVHG-PGANH-100 PROOVHG-TGANH-100 PROOVHG-PGANH-50 PROOVHG-PGANH-500 PROOVHG-TGANH-500
Gallium standard solution, for ICP, in 5% HNO₃, 0,5% HCl, VHG
Standards Kationischer Standards Gallium-Standards
The solutions are intended for use as a certified reference material or calibration standard for inductively coupled plasma optical emission spectroscopy (ICP-OES), inductively coupled plasma mass spectrometry (ICP-MS), flame or furnace atomic absorption spectroscopy (AA or GFAA), x-ray fluorescence spectroscopy (XRF), and other techniques for elemental analysis.
Standards: Traceable to NIST, SRM.