Gadolinium standard solution, for ICP, in 5% HNO₃, VHG
Lieferant: LGC Standards PROMOCHEM
The solutions are intended for use as a certified reference material or calibration standard for inductively coupled plasma optical emission spectroscopy (ICP-OES), inductively coupled plasma mass spectrometry (ICP-MS), flame or furnace atomic absorption spectroscopy (AA or GFAA), x-ray fluorescence spectroscopy (XRF), and other techniques for elemental analysis.
Standards: Traceable to NIST, SRM.