Niobium standard solution, for ICP, in 5% HF, VHG
Lieferant: LGC Standards PROMOCHEM
VHG-PNBF-100 VHG-PNBF-500 VHG-TNBF-100 VHG-TNBF-500
PROOVHG-PNBF-100EA 52 EUR
PROOVHG-PNBF-100 PROOVHG-PNBF-500 PROOVHG-TNBF-100 PROOVHG-TNBF-500
Niobium standard solution, for ICP, in 5% HF, VHG
Standards Kationischer Standards Niob-Standards
The solutions are intended for use as a certified reference material or calibration standard for inductively coupled plasma optical emission spectroscopy (ICP-OES), inductively coupled plasma mass spectrometry (ICP-MS), flame or furnace atomic absorption spectroscopy (AA or GFAA), x-ray fluorescence spectroscopy (XRF), and other techniques for elemental analysis.
Standards: Traceable to NIST, SRM.