Nickel standard solution, for ICP, in 5% HNO₃, VHG
Lieferant: LGC Standards PROMOCHEM
VHG-PNIN-100
VHG-PNIN-500
VHG-PNIN-50
VHG-TNIN-100
VHG-TNIN-500
PROOVHG-PNIN-100EA
50
EUR
PROOVHG-PNIN-100
PROOVHG-PNIN-500
PROOVHG-PNIN-50
PROOVHG-TNIN-100
PROOVHG-TNIN-500
Nickel standard solution, for ICP, in 5% HNO₃, VHG
Standards
Kationischer Standards
Nickel-Standards
The solutions are intended for use as a certified reference material or calibration standard for inductively coupled plasma optical emission spectroscopy (ICP-OES), inductively coupled plasma mass spectrometry (ICP-MS), flame or furnace atomic absorption spectroscopy (AA or GFAA), x-ray fluorescence spectroscopy (XRF), and other techniques for elemental analysis.
Standards: Traceable to NIST, SRM.