Antimony standard solution, for ICP, in 1% HNO₃ and Tartaric Acid, VHG
Lieferant: LGC Standards PROMOCHEM
VHG-PSBWTN-100 VHG-LSBWTN-100 VHG-PSBWTN-250 VHG-PSBWTN-500 VHG-TSBWTN-100 VHG-TSBWTN-500
PROOVHG-PSBWTN-100EA 51 EUR
PROOVHG-PSBWTN-100 PROOVHG-LSBWTN-100 PROOVHG-PSBWTN-250 PROOVHG-PSBWTN-500 PROOVHG-TSBWTN-100 PROOVHG-TSBWTN-500
Antimony standard solution, for ICP, in 1% HNO₃ and Tartaric Acid, VHG
Standards Kationischer Standards Antimon-Standards
The solutions are intended for use as a certified reference material or calibration standard for inductively coupled plasma optical emission spectroscopy (ICP-OES), inductively coupled plasma mass spectrometry (ICP-MS), flame or furnace atomic absorption spectroscopy (AA or GFAA), x-ray fluorescence spectroscopy (XRF), and other techniques for elemental analysis.
Standards: Traceable to NIST, SRM.