Antimony standard solution, for ICP, in 1% HNO₃ and Tartaric Acid, VHG
Lieferant: LGC Standards PROMOCHEM
VHG-PSBWTN-100
VHG-LSBWTN-100
VHG-PSBWTN-250
VHG-PSBWTN-500
VHG-TSBWTN-100
VHG-TSBWTN-500
PROOVHG-PSBWTN-100EA
51
EUR
PROOVHG-PSBWTN-100
PROOVHG-LSBWTN-100
PROOVHG-PSBWTN-250
PROOVHG-PSBWTN-500
PROOVHG-TSBWTN-100
PROOVHG-TSBWTN-500
Antimony standard solution, for ICP, in 1% HNO₃ and Tartaric Acid, VHG
Standards
Kationischer Standards
Antimon-Standards
The solutions are intended for use as a certified reference material or calibration standard for inductively coupled plasma optical emission spectroscopy (ICP-OES), inductively coupled plasma mass spectrometry (ICP-MS), flame or furnace atomic absorption spectroscopy (AA or GFAA), x-ray fluorescence spectroscopy (XRF), and other techniques for elemental analysis.
Standards: Traceable to NIST, SRM.