Sulphur standard solutions for ICP, VHG
Lieferant: LGC Standards PROMOCHEM
VHG-PSW-100 VHG-PSW-500 VHG-TSW-100 VHG-TSW-500
PROOVHG-PSW-100EA 50 EUR
PROOVHG-PSW-100 PROOVHG-PSW-500 PROOVHG-TSW-100 PROOVHG-TSW-500
Sulphur standard solutions for ICP, VHG
Standards Kationischer Standards Schwefel-Standards
This standard solution is intended for use as a certified reference material and calibration standard for inductively coupled plasma optical emission spectroscopy (ICP-OES), inductively coupled plasma mass spectrometry (ICP-MS), flame or furnace atomic absorption spectroscopy (AA or GFAA), x-ray fluorescence spectroscopy (XRF) and other techniques for elemental analysis.
A serial numbered certificate is provided from an ISO/IEC 17025 calibration laboratory to indicate instrument traceability to standards provided by the National Institute of Standards and Technology.
Standards: Traceable to NIST and SRM.