Viscosity standards, VHG
Lieferant: LGC Standards PROMOCHEM
VHG-VISC20-500
VHG-VISC60A
VHG-VISC900
VHG-VISC30-500
VHG-VISC120
VHG-VISC10-500
VHG-VISC50-500
PROOVHG-VISC20-500EA
112
EUR
PROOVHG-VISC20-500
PROOVHG-VISC60A
PROOVHG-VISC900
PROOVHG-VISC30-500
PROOVHG-VISC120
PROOVHG-VISC10-500
PROOVHG-VISC50-500
Viscosity standards, VHG
Standards
Viskosimetrie-Standards
These standard solutions are intended for use as a certified reference materiasl and calibration standards for viscometric instrumentation, using ASTM Method D445 and other viscometric methods.
Standards: Accredited to ISO 9001, ISO 17034 and ISO/IEC 17025. Traceable to NIST.