Elemental analysis standards, PTP sample of oils, VHG
Lieferant: LGC Standards PROMOCHEM
VHG-VPTPMO-25G
PROOVHG-VPTPMO-25GEA 147 EUR
PROOVHG-VPTPMO-25G
Elemental analysis standards, PTP sample of oils, VHG
Standards Mineralölindustrie-Standards
The PTP has been specially designed for analytical laboratories that analyse new and used oils for metals, sulphur, viscosity and particle count.
  • Monitor and improve measurements processes
  • Demonstrate analytical competency
  • Method and instrumentation validation
  • 25 analytes
Standards: ISO/IEC 17043 accredited.