Single element petroleum atandard, calcium in hydrocarbon oil, CRM, VHG
Lieferant: LGC Standards PROMOCHEM
VHGOCA5000-50G
PROOVHGOCA5000-50GEA
92
EUR
PROOVHGOCA5000-50G
Single element petroleum atandard, calcium in hydrocarbon oil, CRM, VHG
Standards
Kationischer Standards
Calcium-Standards
This solution is intended for use as a certified reference material or calibration standard for the analysis of this element in petroleum products or other organic matrices using inductively coupled plasma optical emission spectroscopy (ICP-OES), inductively coupled plasma mass spectrometry (ICP-MS), rotating disc electrode atomic emission spectroscopy (RDE-AES), flame or furnace atomic absorption spectroscopy (AA or GFAA), and other techniques for elemental analysis.
Standards: Traceable to NIST, SRM.