Elemental analysis standards, sulphur standards, VHG
Lieferant: LGC Standards PROMOCHEM
VHGSDSL1500100
VHGSDSL200100
VHGSDSL25100
VHGSDSL2P100
VHGSDSL3000100
VHGSDSL3P100
VHGSDSL400100
VHGSDSL4P100
VHGSDSL5000100
VHGSDSL500100
VHGSDSL5P100
VHGSDSL7500100
VHGSDSL5100
VHGSDSL1000100
VHGSDSL300100
VHGSDSL10100
VHGSDSL15100
VHGSDSL20100
VHGSDSL50100
VHGSDSL75100
VHGSDSL1P100
VHGSDSL100100
VHGSDSL750100
VHGSDSLBLK100
PROOVHGSDSL1500100EA
135
EUR
PROOVHGSDSL1500100
PROOVHGSDSL200100
PROOVHGSDSL25100
PROOVHGSDSL2P100
PROOVHGSDSL3000100
PROOVHGSDSL3P100
PROOVHGSDSL400100
PROOVHGSDSL4P100
PROOVHGSDSL5000100
PROOVHGSDSL500100
PROOVHGSDSL5P100
PROOVHGSDSL7500100
PROOVHGSDSL5100
PROOVHGSDSL1000100
PROOVHGSDSL300100
PROOVHGSDSL10100
PROOVHGSDSL15100
PROOVHGSDSL20100
PROOVHGSDSL50100
PROOVHGSDSL75100
PROOVHGSDSL1P100
PROOVHGSDSL100100
PROOVHGSDSL750100
PROOVHGSDSLBLK100
Elemental analysis standards, sulphur standards, VHG
Standards
Kationischer Standards
Schwefel-Standards
This standard solution is intended for use as a certified reference material and calibration standard for the analysis of sulphur in various petroleum products using x-ray fluorescence spectrometry (XRF), combustion and other techniques for sulphur analysis. It is suitable for use with methods such as ASTM D2622, D4294, D5453, D7039, D7212, D7220 and others.
Standards: Traceable to NIST and SRM.