TCM defined serum-free growth medium
Lieferant: PROTIDE PHARMACEUTICALS
1001
1002
1005
PRPH1001EA
0
EUR
PRPH1001
PRPH1002
PRPH1005
TCM defined serum-free growth medium
Zellkultur-Ergänzungen und -Zusätze
General serum replacement for many cell types from a variety of species. TCM™ is a defined, multi-purpose serum replacement useful in studying the effects of growth factors and the production of proteins. TCM™ contains highly purified, heat-treated bovine serum albumin, heat-treated bovine transferrin and recombinant insulin.
- Defined, serum-free
- No hormones, antibiotics, growth factors, phenol red, Ig
- 50X concentrate
- Can be utilised with a wide variety of basal media
Standards: cGMP manufactured.