Short-term storage solutions, 2-8CELLsius
Lieferant: PROTIDE PHARMACEUTICALS
PP338-100
PP338-500
PP338-1000
PP338-1000B
PRPHPP338-100EA
0
EUR
PRPHPP338-100
PRPHPP338-500
PRPHPP338-1000
PRPHPP338-1000B
Short-term storage solutions, 2-8CELLsius
Zellkultur-Ergänzungen und -Zusätze
Ready to use, balanced, cytoprotective, protein-free solution with bio-stabilisers for short-term storage, regenerative medicine, stem cell processing and transportation of biological material. Short-term storage can be achieved at 2 to 8 °C or at ambient temperature for 24 to 48 hours. Does not contain any growth factors, hormones, or antibiotics. 2-8CELLsius™ can also be utilised as a cell washing solution.
- TYPE II drug master file
- USP grade components/highest grade
- Supports high cell viability
- No animal or human proteins
- No growth factors, hormones or antibiotics
- Ready to use/sterile membrane filtered
Standards: cGMP manufactured.