Cryopreservation solutions, defined, 2-8CELLsius + DMSO
Lieferant: PROTIDE PHARMACEUTICALS
PP338DM5-10
PP338DM5-50
PP338DM5-100
PP338DM5-100B
PP338DM10-10
PP338DM10-50
PP338DM10-100
PP338DM10-100B
338DM10-1000B
PP338DM20-100B
338DM20-1000B
PRPHPP338DM5-10EA
0
EUR
PRPHPP338DM5-10
PRPHPP338DM5-50
PRPHPP338DM5-100
PRPHPP338DM5-100B
PRPHPP338DM10-10
PRPHPP338DM10-50
PRPHPP338DM10-100
PRPHPP338DM10-100B
PRPH338DM10-1000B
PRPHPP338DM20-100B
PRPH338DM20-1000B
Cryopreservation solutions, defined, 2-8CELLsius + DMSO
Zellkultur-Ergänzungen und -Zusätze
When combined with DMSO, 2-8CELLsius™ can be utilised as a cryopreservation solution. 2-8CELLsius™ + DMSO is a cytoprotective, protein-free solution, with bio-stabilisers that support high post thaw cell viability. Optimal recovery is achieved through a novel approach to cell physiology at ultra-low temperatures.
- TYPE II drug master file
- USP grade components/highest grade
- Supports high cell viability
- No animal or human proteins
- No growth factors, hormones, antibiotics
- Ready to use/sterile membrane filtered
Standards: cGMP manufactured.