Human Recombinant Sulfotransferase 1C2 (from E. coli)
Lieferant: ProSci Inc.
92-132
PRSI92-132EA 588 EUR
PRSI92-132
Human Recombinant Sulfotransferase 1C2 (from E. coli)
Enzyme
Sulfotransferase 1C2 (SULT1C2) is a cytosolic enzyme member of the Sulfotransferase 1 family.
This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: N-6 His tag
Human SULT1C2 is primarily expressed in the adult stomach, kidney and thyroid gland, and in the fetal kidney and liver. SULT1C2 catalyzes the sulfate conjugation of drugs, xenobiotic compounds, hormones, and neurotransmitters. SULT1C2 may be involved in the activation of carcinogenic hyroxylamines. It shows activity towards p-nitrophenol and N-hydroxy-2-acetylamino-fluorene (N-OH-2AAF).