Filter for half masks, P940
Lieferant: Portwest
P940GRR
PRTWP940GRREA 47.3 EUR
PRTWP940GRR
Filter for half masks, P940
Atemschutzmasken Mehrweg-Atemschutzmasken Maskenfilter
P940 is a particle filter that protects against dusts. It is equipped with a special thread for pair use only and it can be used only on the P410 half mask.
  • Lightweight and comfortable
  • Screw-in connection
This filter is a P3 R type and protects against toxic substances such as particles and microorganisms.
Standards: EN 143 Type P 3R