Safety spectacles, PW35
Lieferant: Portwest
PW35CLR
PRTWPW35CLREA 5.1 EUR
PRTWPW35CLR
Safety spectacles, PW35
Schutzbrillen
The Portwest safeguard clear lens safety glasses PW35CLR are suited to providing eye protection in a wide range of industries. The glasses are scratch resistant for improved durability, and they feature adjustable arms and seven-position ratchet temples for enhanced fit.
  • Clear lens is ideal for providing eye protection during a range of tasks
  • Combines a polycarbonate lens with a nylon frame
  • Side arms are adjustable in length and feature seven-position ratchet temples
  • Designed to protect against low-energy impact up to 45 metres per second
  • Scratch-resistant coating improves longevity of glasses
The safeguard safety glasses can be adjusted to ensure the wearer can receive the most comfortable secure fit possible. The side arms can be adjusted in length, and the temples feature a seven-position ratchet design to alter the angle of the glasses.
The lenses have been treated with a scratch-resistant coat. This reduces the risk of the glasses becoming damaged when left on table surfaces, in turn reducing the risk of vision becoming impaired by scratches to the lenses.
Standards: EN 166 - Personal Eye Protection