Adapter kit
Lieferant: Cytiva
PSA410
PSA410EA 45.38 EUR
PSA410
Adapter kit
Netzgeräte
Connects 4 mm output jack to 4 mm high voltage leads.
Note that this adaptor kit is not CE compliant.
Bestellinformation: Supplied with Allen wrench.