Quin 2 potassium salt hydrate, Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY

Q0501-10MG
Q0501-10MGEA 553 EUR
Q0501-10MG
Quin 2 potassium salt hydrate, Sigma-Aldrich®
Quin 2 potassium salt hydrate

Calcium-specific chelator and indicator. Upon binding calcium there is a shift in the UV spectrum and an enhancement of fluorescence.

Formel: C₂₆H₂₇N₃O
Lagertemperatur: Kühlschrank
MDL: MFCD00167201
CAS-Nummer: 149022-19-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=acbfaac3c13eb4d19a83a89b2d5d36cdf0482299&vid=e4c080b0a0515da0b5706f56e13d937a241d1fd5&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR