Iodate in Water, certified reference material (CRM), Supelco®
Lieferant: Merck
QC1243-20ML
QC1243-20MLEA
89.2
EUR
QC1243-20ML
Iodate in Water, certified reference material (CRM), Supelco®
Standards
Anionische Standards
Iodate-Standards
Values of analytes vary lot to lot.
Standards: his certified reference material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025:2005.