Multi-element standard, Anions whole volume, certified reference materials (CRMs), for ion chromatography (IC), Supelco®
Lieferant: Merck
QC3364-500ML
QC3364-500MLEA 106 EUR
QC3364-500ML
Multi-element standard, Anions whole volume, certified reference materials (CRMs), for ion chromatography (IC), Supelco®
Standards Anionische Standards Anionische Multi-Element-Standards
Values of analytes vary lot to lot. Some lots may not contain all analytes..
Standards: This certified reference material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025:2005.
Bitte beachten Storage temperature: 2 to 8 °C.