Scheidetrichter, birnenförmig
Lieferant: QVF LABORTECHNIK
ALG0050706
QVFLALG0050706EA 0 EUR
QVFLALG0050706
Scheidetrichter, birnenförmig
Trichter Scheidetrichter
Made of borosilicate glass.
  • Stopcock with glass key
  • Without graduations
  • Excellent chemical and temperature resistance
The separating funnels are used for discontinuous shaking out of solvents.
Standards: Complies to DIN 12 451.