Condenser, jacketed
Lieferant: QVF LABORTECHNIK
GSG0107303
QVFLGSG0107303EA 820 EUR
QVFLGSG0107303
Condenser, jacketed
Kühler
Borosilicate glass 3.3, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes