Natriumchlorid-Lösung 5 M, steril

Lieferant: G Bioscienes

R006
R006EA 86.4 EUR
R006
Natriumchlorid-Lösung 5 M, steril
Natriumchlorid

A sterile solution of 5M sodium chloride prepared with ACS Grade sodium chloride.

Formel: NaCl
MDL: MFCD00003477
CAS-Nummer: 7647-14-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR