Kaliumacetat 3 mol/l (3 N) aqueous solution
Lieferant: G Bioscienes
Synonyme:
Essigsäure Kaliumsalz
R011
R011EA
62.9
EUR
R011
Kaliumacetat 3 mol/l (3 N) aqueous solution
Kaliumacetat
Formel:
CH₃COOK Molecular Weight: 98.14 g/mol Schmelzpunkt: 304 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00012458 CAS-Nummer: 127-08-2 EINECS: 204-822-2 Merck Index: 13,07687 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...