Molekularbiologisches-Kit universal
Lieferant: G Bioscienes
Achtung
R033
R033EA 579 EUR
R033
Molekularbiologisches-Kit universal
Molecular Biology Kit
Molecular biology universal kit, contains 25 ml each of ammonium acetate, calcium chloride, EDTA, lithium acetate, magnesium chloride, potassium acetate, potassium chloride, SDS, sodium chloride, TE, Tris pH 7,0 and Tris pH 8,0
All the reagents in the kit are certified DNase- and RNase-free.
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f18414f2f2530dfdf0edf065728ac8c2f1fff8ad&vid=3a198f3378aaa3d55d7de59c7ee1265443a73531&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9a80b41aae3d45f9440d931003e5299b07279d3a&vid=3644375d8ee47ec22264f175bd0d51c46214843e&prid=web&clid=SAPDAM