Molekularbiologisches-Kit universal
Lieferant: G Bioscienes
|
Achtung
|
R033
R033EA
579
EUR
R033
Molekularbiologisches-Kit universal
Molecular Biology Kit
Molecular biology universal kit, contains 25 ml each of ammonium acetate, calcium chloride, EDTA, lithium acetate, magnesium chloride, potassium acetate, potassium chloride, SDS, sodium chloride, TE, Tris pH 7,0 and Tris pH 8,0
All the reagents in the kit are certified DNase- and RNase-free.