Reactive Red 120, Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY
Gefahr


M-Clarity™ Qualitätsstufe = MQ200

R0378-50G
R0378-50GEA 122 EUR
R0378-50G
Reactive Red 120, Sigma-Aldrich®
Reactive Red 120

Reactive Red 120 is a triazine-based azo dye.

Formel: C₄₄H₂₄Cl₂N₁₄Na₆O₂S₆
Molecular Weight: 1469,98 g/mol
Lagertemperatur: Room Temperature
MDL: MFCD00132157
CAS-Nummer: 61951-82-4
EINECS: 263-351-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c9c6865149c67f6ed48466f7971e802c1d77900c&vid=c9c6865149c67f6ed48466f7971e802c1d77900c&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Appearance (Colour) Red to Brown
Appearance (Form) Powder
Solubility (Colour) Red
Solubility (Form), 1 mg/ml, 50% EtOH Solution
Carbon ≥18.0 %
Nitrogen ≥6.7 %
Sodium (Na) ≥8 %
Dye Content ≥50 %
Recommended Retest Period 6 Years

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR