D(-)-Sorbit solution_SC 1 M

Lieferant: G Bioscienes

Synonyme: D(-)-Glucit, (2R,3R,4R,5S)-(-)-1,2,3,4,5,6-Hexanhexol, D(-)-Sorbitol, D(-)-Glucitol

R038
R038EA 74.2 EUR
R038
D(-)-Sorbit solution_SC 1 M
D(-)-Sorbit

Quality DNase and RNase free reagent solution for yeast research.

Formel: HOH₂C(CH(OH))₄CH₂OH
Molecular Weight: 182.17 g/mol
Siedepunkt: 295
Lagertemperatur: Kühlschrank
MDL: MFCD00004708
CAS-Nummer: 50-70-4
Merck Index: 13,08797
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f18414f2f2530dfdf0edf065728ac8c2f1fff8ad&vid=3a198f3378aaa3d55d7de59c7ee1265443a73531&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR