Ethidiumbromid 0,625 mg/ml aqueous solution

Lieferant: G Bioscienes

Synonyme: 3,8-Diamino-5-ethyl-6-phenylphenanthridiniumbromid

R041
R041EA 139 EUR
R041
Ethidiumbromid 0,625 mg/ml aqueous solution
Ethidiumbromid
Formel: C₂₁H₂₀BrN₃
Molecular Weight: 394.31 g/mol
Siedepunkt: 72
MDL: MFCD00011724
CAS-Nummer: 1239-45-8
Merck Index: 13,04751

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR