Adenosine-5’-Triphosphate (ATP) 100 mM in wässriger Lösung ≥99% für die Molekularbiologie
Lieferant: Thermo Fisher Scientific
R0441
R0441EA
43.72
EUR
R0441
Adenosine-5’-Triphosphate (ATP) 100 mM in wässriger Lösung ≥99% für die Molekularbiologie
Adenosine-5’-Triphosphate (ATP solution)
ATP (adenosine 5'-triphosphate) is an extremely stable nucleotide, and it is supplied as 100 mM aqueous solution titrated to pH 7.3-7.5 with NaOH. This nucleotide is used in different molecular biology applications, and it has greater than 99% purity.
- Greater than 99% purity confirmed by HPLC
- Functionally tested in in vitro transcription
- High stability – stable for two years at -20 °C and after multiple freeze-thaw cycles
Applications: in vitro transcription, aRNA synthesis, siRNA synthesis, RNA amplification, Ligation, Phosphorylation
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...