Wasser Nuclease_free, Fermantas

Lieferant: Thermo Fisher Scientific

Synonyme: Wasserstoffoxid, Hydrogenoxid

R0581 R0582
R0581EA 31.17 EUR
R0581 R0582
Wasser Nuclease_free, Fermantas
Wasser

This nuclease-free water is deionized and 0.22 µM membrane-filtered nuclease-free water


  • Free of endo, exodeoxyribonucleases, ribonucleases, and phosphatases


It is ideal for all molecular biology applications.

Formel: H₂O
Molecular Weight: 18.02 g/mol
Siedepunkt: 100
Schmelzpunkt: 0
Dichte: 1 g/cm³ (4 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00011332
CAS-Nummer: 7732-18-5
EINECS: 231-791-2
Merck Index: 13,10098

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR