Pig Pepsin (from Stomach)

Lieferant: SIGMA ALDRICH MICROSCOPY

R2283-100ML R2283-15ML
R2283-100MLEA 194 EUR
R2283-100ML R2283-15ML
Pig Pepsin (from Stomach)
Enzyme

Pepsin is used for proteolytic digestion of formalin-fixed paraffin-embedded (FFPE) tissue sections prior to application of antibodies. In immunohistochemistry (IHC), most commonly used fixatives such as formalin mask tissue antigens (cellular, membrane, and nuclear) by their intrinsic crosslinking. This masking results in poor or no staining in IHC. Pepsin digestion of FFPE tissue sections improves accessibility of antibodies to tissue antigens. Ready to use pepsin reagent, antigen retriever has been used for antigen unmasking in immunohistochemistry.


  • Antigen Retriever
  • GHS 7

Bestellen

TECHNISCHE DATEN

Enzymname Pepsin
Environmentally Preferable
Ursprung Stomach
Spezies Pig
Konzentration Ready to Use
Lagerbedingungen 2…8 °C
M-Clarity Qualitätsstufe MQ100

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR