Reticulocyte stain, Sigma-Aldrich®
Lieferant: SIGMA ALDRICH MICROSCOPY
R4132-120ML
R4132-120MLEA
164
EUR
R4132-120ML
Reticulocyte stain, Sigma-Aldrich®
Reticulocyte stain
Utilizes New Methylene Blue for the identification of reticulocytes in blood films.